4-(2-Bromoethyl)benzoic acid methyl ester ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B1944-25G B1944-5G
TCIAB1944-25GEA 541 EUR
TCIAB1944-25G TCIAB1944-5G
4-(2-Bromoethyl)benzoic acid methyl ester ≥98.0% (by titrimetric analysis)
4-(2-Bromoethyl)benzoic acid methyl ester
Formule: C₉H₉BrO₂
MDL Nummer: MFCD00016541
CAS nummer: 52062-92-7
EINECS: 257-632-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce79a05ef13861acf78337691a2b855ca3a00936&vid=ce79a05ef13861acf78337691a2b855ca3a00936&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 98.0 %
Solubility in hot Methanol almost transparency

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR