6-Bromoquinoline ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B2015-1G B2015-5G
TCIAB2015-1GEA 29.7 EUR
TCIAB2015-1G TCIAB2015-5G
6-Bromoquinoline ≥95.0% (by GC)
6-Bromoquinoline
Formule: C₉H₆BrN
Kookpunt: 116°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00024023
CAS nummer: 5332-25-2
EINECS: 226-238-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9de49f5ade769d3a32d92c18f20f9dcc1bcd3b2a&vid=9dc8762f87571770a44f65d64f2997b9261e3979&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White or Colorless to Yellow powder to lump to clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR