1,4-Bis(2-chloroethyl)benzene ≥96.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B2201-1G
TCIAB2201-1GEA 160 EUR
TCIAB2201-1G
1,4-Bis(2-chloroethyl)benzene ≥96.0% (by GC)
1,4-Bis(2-chloroethyl)benzene
Formule: C₁₀H₁₂Cl₂
CAS nummer: 7379-84-2

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 96.0 %
Melting point 44.0 to 48.0 deg-C

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR