2-Bromo-4-chloroaniline ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B2268-25G B2268-5G
TCIAB2268-25GEA 121 EUR
TCIAB2268-25G TCIAB2268-5G
2-Bromo-4-chloroaniline ≥98.0% (by GC)
2-Bromo-4-chloroaniline
Formule: C₆H₅BrClN
MW: 206.47 g/mol
Kookpunt: 127°C
Smeltpunt: 69
MDL Nummer: MFCD00041313
CAS nummer: 873-38-1
EINECS: 212-837-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c642cfd52ab2b1ccac287844b2de2f53d610c9a5&vid=90f7360eb324904ab0634d219df9377162af7d1f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 64.0 to 68.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR