Methyl-4-bromobutyrate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B2308-25G B2308-250G
TCIAB2308-25GEA 60.5 EUR
TCIAB2308-25G TCIAB2308-250G
Methyl-4-bromobutyrate ≥98.0% (by GC)
Methyl-4-bromobutyrate
Formule: C₅H₉BrO₂
MDL Nummer: MFCD00041482
CAS nummer: 4897-84-1
EINECS: 225-523-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5875f00eec1563be2be0428e45714fc9ab74fb74&vid=5875f00eec1563be2be0428e45714fc9ab74fb74&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR