Benzaldehyde ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B2379-500G
TCIAB2379-500GEA 38.5 EUR
TCIAB2379-500G
Benzaldehyde ≥98.0% (by GC)
Benzaldehyde
Formule: C₆H₅CHO
Kookpunt: 179°C
Smeltpunt: -56°C
Dichtheid: 1,049 g/cm³ (20 °C)
Vlampunt: 62°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003299
CAS nummer: 100-52-7
EINECS: 202-860-4
UN: 1990
ADR: 9,III
Merck Index: 14,01058
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d08b9d4df5f89af4acb7445420ff527101170a42&vid=d08b9d4df5f89af4acb7445420ff527101170a42&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR