1-Benzyl-N-(tert-butoxycarbonyl)-L-aspartate ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

B2408-25G B2408-5G
TCIAB2408-25GEA 179 EUR
TCIAB2408-25G TCIAB2408-5G
1-Benzyl-N-(tert-butoxycarbonyl)-L-aspartate ≥98.0% (by HPLC, titration analysis)
1-Benzyl-N-(tert-butoxycarbonyl)-L-aspartate
Formule: C₁₆H₂₁NO₆
MDL Nummer: MFCD00065563
CAS nummer: 30925-18-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 97.0 to 101.0 deg-C
Specific rotation [a]20/D -21.0 to -26.0 deg (C=1, MeOH)

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR