Bis(2,2,2-trifluoroethyl)phosphite ≥94.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B2627-25G B2627-5G
TCIAB2627-25GEA 139 EUR
TCIAB2627-25G TCIAB2627-5G
Bis(2,2,2-trifluoroethyl)phosphite ≥94.0% (by GC)
Bis(2,2,2-trifluoroethyl)phosphite
Formule: C₄H₅F₆O₃P
Kookpunt: 44°C
Vlampunt: 76°C
MDL Nummer: MFCD00063314
CAS nummer: 92466-70-1

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 94.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR