tert-Butyl (diethylphosphono)acetate ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: tert-Butyl 2-(diethoxyphosphoryl)acetate, Diethyl(2-tert-butoxycarbonylmethyl)phosphonate

B2814-1G B2814-5G
TCIAB2814-1GEA 25.3 EUR
TCIAB2814-1G TCIAB2814-5G
tert-Butyl (diethylphosphono)acetate ≥97.0% (by GC)
tert-Butyl (diethylphosphono)acetate
Formule: C₁₀H₂₁O₅P
Kookpunt: 152°C
MDL Nummer: MFCD00075414
CAS nummer: 27784-76-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea5fb6ec8766dcde1519032f136581a40108093&vid=bea5fb6ec8766dcde1519032f136581a40108093&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR