4-Bromobenzaldehyde diethyl acetal ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 4-Bromo-α,α-diethoxytoluene, 1-Bromo-4-(diethoxymethyl)benzene

B2845-25G
TCIAB2845-25GEA 69.3 EUR
TCIAB2845-25G
4-Bromobenzaldehyde diethyl acetal ≥97.0% (by GC)
4-Bromobenzaldehyde diethyl acetal
Formule: C₁₁H₁₅BrO₂
MW: 259.14 g/mol
Vlampunt: 105°C
MDL Nummer: MFCD01863514
CAS nummer: 34421-94-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=835e5e41c7669113445e93660d0b74641b8b7b9a&vid=835e5e41c7669113445e93660d0b74641b8b7b9a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR