4-Bromo-α,α,α,2-tetrafluorotoluene ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B2946-5G B2946-25G
TCIAB2946-5GEA 31.9 EUR
TCIAB2946-5G TCIAB2946-25G
4-Bromo-α,α,α,2-tetrafluorotoluene ≥98.0% (by GC)
4-Bromo-α,α,α,2-tetrafluorotoluene
Formule: C₇H₃BrF₄
MDL Nummer: MFCD01075254
CAS nummer: 142808-15-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d63fd10449ab2916122249d9bc91027c0ae7a978&vid=d63fd10449ab2916122249d9bc91027c0ae7a978&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR