4-Butylbenzyl bromide ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

B3005-1G B3005-5G
TCIAB3005-1GEA 72.6 EUR
TCIAB3005-1G TCIAB3005-5G
4-Butylbenzyl bromide ≥97.0% (by GC)
4-Butylbenzyl bromide
Formule: C₁₁H₁₅Br
Kookpunt: 144°C
CAS nummer: 10531-16-5
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR