N-(tert-Butoxycarbonyl)-D-leucine hydrate ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

B3007-25G B3007-5G
TCIAB3007-25GEA 102 EUR
TCIAB3007-25G TCIAB3007-5G
N-(tert-Butoxycarbonyl)-D-leucine hydrate ≥98.0% (by titrimetric analysis)
N-(tert-Butoxycarbonyl)-D-leucine hydrate
Formule: C₁₁H₂₁NO₄
MDL Nummer: MFCD00038294
CAS nummer: 16937-99-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 % (calcd.on anh.substance)
Melting point 85.0 to 89.0 deg-C
Specific rotation [a]20/D +22.0 to +28.0 deg (C=2, AcOH)
Water 5.0 to 9.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR