(R)-(-)-1-Boc-3-hydroxypyrrolidine ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B3054-1G B3054-5G
TCIAB3054-1GEA 25.3 EUR
TCIAB3054-1G TCIAB3054-5G
(R)-(-)-1-Boc-3-hydroxypyrrolidine ≥98.0% (by GC)
(R)-(-)-1-Boc-3-hydroxypyrrolidine
Formule: C₉H₁₇NO₃
MDL Nummer: MFCD01317838
CAS nummer: 109431-87-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 60.0 to 65.0 deg-C
Specific rotation [a]20/D -25.0 to -29.0 deg (C=1, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR