1-Bromo-5-fluoro-2-nitrobenzene ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B3061-5G
TCIAB3061-5GEA 58.3 EUR
TCIAB3061-5G
1-Bromo-5-fluoro-2-nitrobenzene ≥98.0% (by GC)
1-Bromo-5-fluoro-2-nitrobenzene
Formule: C₆H₃BrFNO₂
MDL Nummer: MFCD00792441
CAS nummer: 700-36-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd6e8d23d5552829079a27250be8d2bfb4473aef&vid=cd6e8d23d5552829079a27250be8d2bfb4473aef&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 41.0 to 45.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR