N-ɑ-Fmoc-N-ε-boc-L-lysine ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

B3072-25G B3072-5G
TCIAB3072-25GEA 101 EUR
TCIAB3072-25G TCIAB3072-5G
N-ɑ-Fmoc-N-ε-boc-L-lysine ≥98.0% (by HPLC, titration analysis)
N-alpha-Fmoc-N-epsilon-boc-L-lysine
Formule: C₂₆H₃₂N₂O₆
Smeltpunt: 124°C
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00037138
CAS nummer: 71989-26-9
EINECS: 276-256-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D -10.0 to -14.0 deg (C=1, DMF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR