Bromo[(2,6-pyridinediyl)bis(3-methyl-1-imidazolyl-2-ylidene)]nickel bromide ≥98.0% (by total nitrogen and titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

B3354-1G
TCIAB3354-1GEA 175 EUR
TCIAB3354-1G
Bromo[(2,6-pyridinediyl)bis(3-methyl-1-imidazolyl-2-ylidene)]nickel bromide ≥98.0% (by total nitrogen and titration analysis)
Bromo[(2,6-pyridinediyl)bis(3-methyl-1-imidazolyl-2-ylidene)]nickel bromide
Formule: C₁₃H₁₃Br₂N₅Ni
CAS nummer: 894102-11-5

Order Now

Specification Test Results

Appearance Orange to Amber to Dark red powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Chelometric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR