5-Bromo-2-methoxybenzyl alcohol ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

B3369-25G B3369-5G
TCIAB3369-25GEA 240 EUR
TCIAB3369-25G TCIAB3369-5G
5-Bromo-2-methoxybenzyl alcohol ≥98.0% (by GC)
5-Bromo-2-methoxybenzyl alcohol
Formule: C₈H₉BrO₂
MDL Nummer: MFCD00004630
CAS nummer: 80866-82-6
EINECS: 279-586-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=063a661dad0962b574881894b5c6b426489120d7&vid=063a661dad0962b574881894b5c6b426489120d7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 71.0 to 75.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR