4-Bromo-2-methylthiophene ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

B3385-25G B3385-5G
TCIAB3385-25GEA 204 EUR
TCIAB3385-25G TCIAB3385-5G
4-Bromo-2-methylthiophene ≥98.0% (by GC)
4-Bromo-2-methylthiophene
Formule: C₅H₅BrS
Kookpunt: 62°C
Vlampunt: 68°C
MDL Nummer: MFCD04116367
CAS nummer: 29421-92-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd678eb7e1efb123c30587aaa41a206d97779f9c&vid=bd678eb7e1efb123c30587aaa41a206d97779f9c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR