Bis(2,4-pentanedionato)bis(2-propanolato)titanium(IV) 75% in isopropanol (IPA)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

B3395-25G B3395-500G
TCIAB3395-25GEA 17.6 EUR
TCIAB3395-25G TCIAB3395-500G
Bis(2,4-pentanedionato)bis(2-propanolato)titanium(IV) 75% in isopropanol (IPA)
Bis(2,4-pentanedionato)bis(2-propanolato)titanium(IV)
Formule: C₁₆H₂₈O₆Ti
Vlampunt: 18°C
MDL Nummer: MFCD00000031
CAS nummer: 17927-72-9
EINECS: 241-866-1
UN: 1219
ADR: 3,II

Order Now

Specification Test Results

Appearance White to Yellow to Orange clear liquid
Concentration (Gravimetric Method) 75.0 to 80.0 w/w%

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR