2-Bromo-4-fluorobenzyl alcohol ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 2-Bromo-4-fluorobenzyl Alcohol, (2-Bromo-4-fluorophenyl)methanol

B3417-5G B3417-25G
TCIAB3417-5GEA 52.8 EUR
TCIAB3417-5G TCIAB3417-25G
2-Bromo-4-fluorobenzyl alcohol ≥98.0% (by GC)
2-Bromo-4-fluorobenzyl alcohol
Formule: C₇H₆BrFO
MW: 205.03
MDL Nummer: MFCD00672925
CAS nummer: 229027-89-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=51c775271123782cc0e180506064fd30a27bfafb&vid=51c775271123782cc0e180506064fd30a27bfafb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 68.0 to 72.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR