Bis(zinc porphyrin) ∼5 µmol/L in dichloromethane for cd spectroscopy

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

B3515-25ML
TCIAB3515-25MLEA 112 EUR
TCIAB3515-25ML
Bis(zinc porphyrin) ∼5 µmol/L in dichloromethane for cd spectroscopy
Bis(zinc porphyrin)
Formule: C₇₄H₉₀N₈Zn₂
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD03459124
CAS nummer: 92995-45-4
UN: 1593
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=91cac7532add3b23173880670b1c0620d6a066c0&vid=d15f059f4b84e570de240608a189277ee25457eb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Yellow to Amber to Dark red clear liquid
Concentration (UV Methoed)) 5.0 to 7.0 Micro_mol/L


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR