Benzoyl trifluoromethanesulphonate ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: Benzoyl Triflate, Trifluoromethanesulphonic acid Benzoyl Ester, Benzoic trifluoromethanesulphonic anhydride

B3567-5G
TCIAB3567-5GEA 503 EUR
TCIAB3567-5G
Benzoyl trifluoromethanesulphonate ≥98.0% (by titrimetric analysis)
Benzoyl trifluoromethanesulphonate
Formule: C₈H₅F₃O₄S
MW: 254.18 g/mol
Kookpunt: 94°C
Vlampunt: 114°C
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD17013536
CAS nummer: 36967-85-8

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR