tert-Butyl phenyl carbonate ≥96.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

B3590-25G B3590-5G
TCIAB3590-25GEA 81.4 EUR
TCIAB3590-25G TCIAB3590-5G
tert-Butyl phenyl carbonate ≥96.0% (by HPLC)
tert-Butyl phenyl carbonate
Formule: C₁₁H₁₄O₃
Kookpunt: 78°C
Vlampunt: 101°C
MDL Nummer: MFCD00008804
CAS nummer: 6627-89-0
EINECS: 229-601-8

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (HPLC) min. 96.0 area%

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR