Bis(phenylacetyl)disulphide ≥98.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

B3623-5G B3623-25G
TCIAB3623-5GEA 38.5 EUR
TCIAB3623-5G TCIAB3623-25G
Bis(phenylacetyl)disulphide ≥98.0% (by HPLC)
Bis(phenylacetyl)disulphide
Formule: C₁₆H₁₄O₂S₂
MDL Nummer: MFCD00513572
CAS nummer: 15088-78-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 59.0 to 62.0 deg-C

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR