4-Bromo-3-fluorophenol ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B3688-25G B3688-5G
TCIAB3688-25GEA 372 EUR
TCIAB3688-25G TCIAB3688-5G
4-Bromo-3-fluorophenol ≥98.0% (by GC, titration analysis)
4-Bromo-3-fluorophenol
Formule: C₆H₄BrFO
Kookpunt: 96°C
MDL Nummer: MFCD00051907
CAS nummer: 121219-03-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca8f6331145ee9a5f7ca619e765cd29c97451f75&vid=ca8f6331145ee9a5f7ca619e765cd29c97451f75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 71.0 to 75.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR