Nɑ-tert-Butoxycarbonyl-N-γ-trityl-L-asparagine ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

B3789-25G B3789-5G
TCIAB3789-25GEA 277 EUR
TCIAB3789-25G TCIAB3789-5G
Nɑ-tert-Butoxycarbonyl-N-γ-trityl-L-asparagine ≥98.0% (by HPLC, titration analysis)
Nalpha-tert-Butoxycarbonyl-N-gamma-trityl-L-asparagine
Formule: C₂₈H₃₀N₂O₅
Smeltpunt: 202°C
MDL Nummer: MFCD00153299
CAS nummer: 132388-68-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D -18.0 to -22.0 deg (C=1, methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR