5-Bromo-2-(methylthio)pyrimidine-4-carboxylic acid ≥97.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B3829-1G B3829-5G
TCIAB3829-1GEA 41.8 EUR
TCIAB3829-1G TCIAB3829-5G
5-Bromo-2-(methylthio)pyrimidine-4-carboxylic acid ≥97.0% (by titrimetric analysis)
5-Bromo-2-(methylthio)pyrimidine-4-carboxylic acid
Formule: C₆H₅BrN₂O₂S
Smeltpunt: 177°C
MDL Nummer: MFCD00238866
CAS nummer: 50593-92-5

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR