2-Bromo-5-hydroxybenzaldehyde ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

B3997-1G B3997-5G
TCIAB3997-1GEA 35.2 EUR
TCIAB3997-1G TCIAB3997-5G
2-Bromo-5-hydroxybenzaldehyde ≥97.0% (by GC)
2-Bromo-5-hydroxybenzaldehyde
Formule: C₇H₅BrO₂
MW: 201.02 g/mol
MDL Nummer: MFCD03033065
CAS nummer: 2973-80-0

Order Now

Specification Test Results

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 97.0 %
Melting point 131.0 to 135.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR