SB-431542 ≥97.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

B4003-100MG B4003-25MG
TCIAB4003-100MGEA 535 EUR
TCIAB4003-100MG TCIAB4003-25MG
SB-431542 ≥97.0% (by HPLC)
SB-431542
Formule: C₂₂H₁₈N₄O₃
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD11045982
CAS nummer: 301836-41-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a2569b7c8db4889023c4ba9830126df10c3f7f24&vid=6ff435dfe9d6aac30b9419c82eb7eaa97d77e886&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 97.0 area%
Water max. 3.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR