N-(tert-Butoxycarbonyl)-L-propargylglycine ≥97.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

B4007-1G
TCIAB4007-1GEA 145 EUR
TCIAB4007-1G
N-(tert-Butoxycarbonyl)-L-propargylglycine ≥97.0% (by HPLC, titration analysis)
N-(tert-Butoxycarbonyl)-L-propargylglycine
Formule: C₁₀H₁₅NO₄
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD01320855
CAS nummer: 63039-48-5

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Neutralization titration) min. 97.0 %
Melting point 88.0 to 92.0 deg-C
Specific rotation [a]20/D +24.0 to +27.0 deg (C=1、MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR