1-Butyl-2,3-dimethylimidazoliumtrifluoromethanesulphonate ≥98.0% (by HPLC, total nitrogen)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B4182-5G B4182-25G
TCIAB4182-5GEA 83.6 EUR
TCIAB4182-5G TCIAB4182-25G
1-Butyl-2,3-dimethylimidazoliumtrifluoromethanesulphonate ≥98.0% (by HPLC, total nitrogen)
1-Butyl-2,3-dimethylimidazoliumtrifluoromethanesulphonate
Formule: C₁₀H₁₇F₃N₂O₃S
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD03427624
CAS nummer: 765910-73-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 42.0 to 46.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR