1-Bromo-2,4-bis(trifluoromethyl)benzene ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B4207-1G B4207-5G
TCIAB4207-1GEA 24.2 EUR
TCIAB4207-1G TCIAB4207-5G
1-Bromo-2,4-bis(trifluoromethyl)benzene ≥97.0% (by GC)
1-Bromo-2,4-bis(trifluoromethyl)benzene
Formule: C₈H₃BrF₆
Kookpunt: 51°C
Vlampunt: 69°C
MDL Nummer: MFCD00074904
CAS nummer: 327-75-3

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR