Butyl valerate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B4322-25ML
TCIAB4322-25MLEA 48.4 EUR
TCIAB4322-25ML
Butyl valerate ≥98.0% (by GC)
Butyl valerate
Formule: C₉H₁₈O₂
Kookpunt: 185°C
Vlampunt: 67°C
MDL Nummer: MFCD00015258
CAS nummer: 591-68-4
EINECS: 209-728-5

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR