2-Bromocarbazole ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B4381-5G B4381-25G
TCIAB4381-5GEA 114 EUR
TCIAB4381-5G TCIAB4381-25G
2-Bromocarbazole ≥98.0% (by GC)
2-Bromocarbazole
Formule: C₁₂H₈BrN
MDL Nummer: MFCD09750430
CAS nummer: 3652-90-2

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 253.0 to 257.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR