3-Bromo-6-chloro-2-methylpyridine ≥97.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B4405-5G
TCIAB4405-5GEA 146 EUR
TCIAB4405-5G
3-Bromo-6-chloro-2-methylpyridine ≥97.0% (by GC)
3-Bromo-6-chloro-2-methylpyridine
Formule: C₆H₅BrClN
Kookpunt: 84°C
Smeltpunt: 47°C
MDL Nummer: MFCD07375117
CAS nummer: 132606-40-7

Order Now

Specification Test Results

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 97.0 %

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR