2,4-Bis(benzyloxy)-6-chloro-1,3,5-triazine ≥95.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B4587-200MG B4587-1G
TCIAB4587-200MGEA 49.5 EUR
TCIAB4587-200MG TCIAB4587-1G
2,4-Bis(benzyloxy)-6-chloro-1,3,5-triazine ≥95.0% (by HPLC, titration analysis)
2,4-Bis(benzyloxy)-6-chloro-1,3,5-triazine
Formule: C₁₇H₁₄ClN₃O₂
CAS nummer: 851030-18-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (Argentmetric Titration) min. 95.0 %
Melting point 64.0 to 68.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR