5-Bromo-2-chloro-4-methylpyridine ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B4626-5G B4626-25G
TCIAB4626-5GEA 28.6 EUR
TCIAB4626-5G TCIAB4626-25G
5-Bromo-2-chloro-4-methylpyridine ≥98.0% (by GC)
5-Bromo-2-chloro-4-methylpyridine
Formule: C₆H₅BrClN
MDL Nummer: MFCD03427663
CAS nummer: 778611-64-6

Order Now

Specification Test Results

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR