2-Bromo-4-fluoropyridine ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B4760-1G B4760-5G
TCIAB4760-1GEA 50.6 EUR
TCIAB4760-1G TCIAB4760-5G
2-Bromo-4-fluoropyridine ≥98.0% (by GC)
2-Bromo-4-fluoropyridine
Formule: C₅H₃BrFN
Vlampunt: 76°C
MDL Nummer: MFCD09260909
CAS nummer: 357927-50-5

Order Now

Specification Test Results

Appearance White to Almost white clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR