4-Bromo-3-methyl-5-(trifluoromethyl)-1H-pyrazole ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B4770-1G
TCIAB4770-1GEA 73.7 EUR
TCIAB4770-1G
4-Bromo-3-methyl-5-(trifluoromethyl)-1H-pyrazole ≥98.0% (by GC)
4-Bromo-3-methyl-5-(trifluoromethyl)-1H-pyrazole
Formule: C₅H₄BrF₃N₂
MDL Nummer: MFCD08060951
CAS nummer: 60061-68-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6a5a2622348a6473a7c51533d183f03bb7cea9&vid=f43b2be9a51dae4ebbcba25636e33f05cb8b05ae&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 105.0 to 109.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR