4-Butyl-4'-cyanobiphenyl ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B4923-5G B4923-25G
TCIAB4923-5GEA 51.7 EUR
TCIAB4923-5G TCIAB4923-25G
4-Butyl-4'-cyanobiphenyl ≥98.0% (by GC)
4-Butyl-4'-cyanobiphenyl
Formule: C₁₇H₁₇N
Kookpunt: 195°C
MDL Nummer: MFCD00506009
CAS nummer: 52709-83-8
EINECS: 258-119-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 47.0 to 51.0 deg-C
Mesomorphic range 45.0 to 51.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR