5-Bromo-2-methylbenzaldehyde ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B5036-1G B5036-5G
TCIAB5036-1GEA 57.2 EUR
TCIAB5036-1G TCIAB5036-5G
5-Bromo-2-methylbenzaldehyde ≥95.0% (by GC)
5-Bromo-2-methylbenzaldehyde
Formule: C₈H₇BrO
MW: 199.05
Kookpunt: 172°C
MDL Nummer: MFCD05662390
CAS nummer: 90050-59-2

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to lump
Purity (GC) min. 95.0 %
Melting point 45.0 to 49.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR