2-Bromo-6-hydrazinylpyridine ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B5120-1G B5120-5G
TCIAB5120-1GEA 35.2 EUR
TCIAB5120-1G TCIAB5120-5G
2-Bromo-6-hydrazinylpyridine ≥98.0% (by GC, titration analysis)
2-Bromo-6-hydrazinylpyridine
Formule: C₅H₆BrN₃
MDL Nummer: MFCD09743754
CAS nummer: 26944-71-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4271fb21fa8cdfa54a75a56e29e00bc82a34570a&vid=4271fb21fa8cdfa54a75a56e29e00bc82a34570a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Potassium iodate Method) min. 98.0 %
Melting point 118.0 to 122.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR