2-Bromoisonicotinonitrile ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

B5189-1G B5189-5G
TCIAB5189-1GEA 55 EUR
TCIAB5189-1G TCIAB5189-5G
2-Bromoisonicotinonitrile ≥98.0% (by GC)
2-Bromoisonicotinonitrile
Formule: C₆H₃BrN₂
MDL Nummer: MFCD07367879
CAS nummer: 10386-27-3
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 77.0 to 81.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR