3-Bromo-2-hydroxybenzonitrile ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B5273-1G B5273-5G
TCIAB5273-1GEA 48.4 EUR
TCIAB5273-1G TCIAB5273-5G
3-Bromo-2-hydroxybenzonitrile ≥98.0% (by GC)
3-Bromo-2-hydroxybenzonitrile
Formule: C₇H₄BrNO
MDL Nummer: MFCD11042854
CAS nummer: 13073-28-4

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 120.0 to 124.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR