3-Bromo-4-iodotoluene ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B5284-5G B5284-25G
TCIAB5284-5GEA 22 EUR
TCIAB5284-5G TCIAB5284-25G
3-Bromo-4-iodotoluene ≥98.0% (by GC)
3-Bromo-4-iodotoluene
Formule: C₇H₆BrI
Kookpunt: 267°C
MDL Nummer: MFCD00079718
CAS nummer: 71838-16-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=84e0e554c930c96317f449f788b9906229e37bed&vid=84e0e554c930c96317f449f788b9906229e37bed&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR