1-Benzyl-3-methylimidazolium hexafluorophosphate ≥98.0% (by HPLC, total nitrogen)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B5726-5G B5726-25G
TCIAB5726-5GEA 35.2 EUR
TCIAB5726-5G TCIAB5726-25G
1-Benzyl-3-methylimidazolium hexafluorophosphate ≥98.0% (by HPLC, total nitrogen)
1-Benzyl-3-methylimidazolium hexafluorophosphate
Formule: C₁₁H₁₃F₆N₂P
MDL Nummer: MFCD07784448
CAS nummer: 433337-11-2

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 130.0 to 134.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR