Boc-L-His(Bzl)-OH ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

B5735-1G B5735-5G
TCIAB5735-1GEA 56.1 EUR
TCIAB5735-1G TCIAB5735-5G
Boc-L-His(Bzl)-OH ≥98.0% (by titrimetric analysis)
Boc-L-His(Bzl)-OH
Formule: C₁₈H₂₃N₃O₄
Smeltpunt: 182°C
MDL Nummer: MFCD00066093
CAS nummer: 20898-44-6
EINECS: 244-105-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Specific rotation [a]20/D +22.0 to +25.0 deg (C=1, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR