PEG5-carboxylic acid tert-butyl ester ≥94.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

B5901-1G B5901-5G
TCIAB5901-1GEA 107 EUR
TCIAB5901-1G TCIAB5901-5G
PEG5-carboxylic acid tert-butyl ester ≥94.0% (by GC)
PEG5-carboxylic acid tert-butyl ester
Formule: C₁₅H₃₀O₇
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD11041117
CAS nummer: 518044-32-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3254a993b461c04562adc36489dc0660d1a87224&vid=5a5f2e831e12c23b79cfbd8a97c915512a71fed1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR