[3,5-Bis(ethoxycarbonyl)phenyl]boronic acid (contains varying amounts of Anhydride)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

B6032-200MG B6032-1G
TCIAB6032-200MGEA 33 EUR
TCIAB6032-200MG TCIAB6032-1G
[3,5-Bis(ethoxycarbonyl)phenyl]boronic acid (contains varying amounts of Anhydride)
[3,5-Bis(ethoxycarbonyl)phenyl]boronic acid
Formule: C₁₂H₁₅BO₆
MDL Nummer: MFCD11867795
CAS nummer: 932378-94-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 and le 107.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR